Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scara5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135855.1 | NP_001129327.1 | 1 - 2 | 330 | 97 | |
XM_006252204.2 | XP_006252266.1 | 1 - 2 | 368 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |