Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108071.1 | NP_001101541.1 | 6 - 7 | 618 | 66 | |
XM_006240915.2 | XP_006240977.1 | 8 - 9 | 1547 | 66 | ||
XM_006240916.2 | XP_006240978.1 | 8 - 9 | 1537 | 66 | ||
XM_006240917.2 | XP_006240979.1 | 8 - 9 | 1544 | 66 | ||
XM_006240918.2 | XP_006240980.1 | 8 - 9 | 1487 | 66 | ||
XM_006240919.2 | XP_006240981.1 | 8 - 9 | 1546 | 66 | ||
XM_006240920.2 | XP_006240982.1 | 7 - 8 | 1376 | 66 | ||
GenBank mRNA | FQ224249.1 | - | 8 - 9 | 847 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |