Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024264.1 | NP_001019435.1 | 4 - 5 | 608 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |