Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039849.2 | NP_001034938.1 | 5 - 6 | 676 | 82 | |
NM_017165.2 | NP_058861.3 | 5 - 6 | 562 | 82 | ||
GenBank mRNA | AB072798.1 | - | 5 - 6 | 676 | 82 | |
AF274028.1 | - | 5 - 6 | 685 | 82 | ||
AY570513.1 | - | 5 - 6 | 664 | 82 | ||
FQ210610.1 | - | 5 - 6 | 470 | 82 | ||
FQ221663.1 | - | 5 - 6 | 492 | 82 | ||
FQ221760.1 | - | 5 - 6 | 470 | 82 | ||
FQ221899.1 | - | 5 - 6 | 470 | 82 | ||
FQ221976.1 | - | 5 - 6 | 473 | 82 | ||
FQ222299.1 | - | 5 - 6 | 470 | 82 | ||
FQ230991.1 | - | 5 - 6 | 458 | 82 | ||
FQ231108.1 | - | 5 - 6 | 528 | 82 | ||
FQ233099.1 | - | 5 - 6 | 470 | 82 | ||
FQ234630.1 | - | 5 - 6 | 470 | 82 | ||
L24896.1 | - | 5 - 6 | 439 | 82 | ||
U37427.1 | - | 5 - 6 | 556 | 82 | ||
X82679.1 | - | 5 - 6 | 449 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |