Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100901.1 | NP_001094371.1 | 2 - 3 | 590 | 83 | |
XM_006235326.2 | XP_006235388.1 | 2 - 3 | 629 | 83 | ||
GenBank mRNA | BC169101.1 | - | 2 - 3 | 590 | 5599966 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |