Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001074561.4 | XP_001074561.2 | 11 - 12 | 1327 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |