Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam83c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749599.3 | XP_003749647.1 | 1 - 2 | 678 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |