Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031026.1 | NP_112288.1 | 7 - 8 | 935 | 98 | |
XM_006255053.2 | XP_006255115.1 | 7 - 8 | 1149 | 98 | ||
XM_006255054.2 | XP_006255116.1 | 7 - 8 | 1149 | 98 | ||
GenBank mRNA | AB008521.1 | - | 2 - 3 | 201 | 98 | |
AB008522.1 | - | 2 - 3 | 201 | 98 | ||
AB008523.1 | - | 2 - 3 | 201 | 98 | ||
AB008524.1 | - | 2 - 3 | 201 | 98 | ||
BC089794.1 | - | 7 - 8 | 956 | 7115029 | 98 | |
U15138.1 | - | 7 - 8 | 935 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |