Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039004.1 | NP_001034093.1 | 2 - 3 | 404 | 104 | |
XM_006255132.2 | XP_006255194.1 | 2 - 3 | 1037 | 104 | ||
GenBank mRNA | BC098748.1 | - | 2 - 3 | 404 | 7365565 | 104 |
BC168149.1 | - | 2 - 3 | 1043 | 7457021 | 104 | |
FQ213930.1 | - | 2 - 3 | 447 | 104 | ||
FQ229498.1 | - | 2 - 3 | 445 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |