Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772250.1 | XP_008770472.1 | 13 - 14 | 1791 | 149 | |
XM_008772251.1 | XP_008770473.1 | 13 - 14 | 1712 | 149 | ||
XM_008772252.1 | XP_008770474.1 | 13 - 14 | 1999 | 149 | ||
XM_008772253.1 | XP_008770475.1 | 12 - 13 | 1870 | 149 | ||
XM_008772254.1 | XP_008770476.1 | 13 - 14 | 2002 | 149 | ||
XM_008772255.1 | XP_008770477.1 | 12 - 13 | 1849 | 149 | ||
XM_008772256.1 | XP_008770478.1 | 12 - 13 | 1846 | 149 | ||
XM_008772257.1 | XP_008770479.1 | 13 - 14 | 2002 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |