Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772250.1 | XP_008770472.1 | 16 - 17 | 2108 | 115 | |
XM_008772251.1 | XP_008770473.1 | 16 - 17 | 2029 | 115 | ||
XM_008772252.1 | XP_008770474.1 | 16 - 17 | 2316 | 115 | ||
XM_008772253.1 | XP_008770475.1 | 15 - 16 | 2187 | 115 | ||
XM_008772254.1 | XP_008770476.1 | 15 - 16 | 2169 | 115 | ||
XM_008772255.1 | XP_008770477.1 | 15 - 16 | 2166 | 115 | ||
XM_008772256.1 | XP_008770478.1 | 15 - 16 | 2163 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |