Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003705.2 | NP_001003705.1 | 5 - 6 | 897 | 72 | |
XM_006255067.2 | XP_006255129.1 | 7 - 8 | 1044 | 72 | ||
GenBank mRNA | AY294328.1 | - | 6 - 7 | 992 | 72 | |
BC086369.1 | - | 5 - 6 | 897 | 7109456 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |