Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108438.2 | NP_001101908.1 | 6 - 7 | 888 | 109 | |
XM_008772314.1 | XP_008770536.1 | 5 - 6 | 829 | 109 | ||
XM_008772315.1 | XP_008770537.1 | 6 - 7 | 921 | 109 | ||
XM_008772316.1 | XP_008770538.1 | 6 - 7 | 918 | 109 | ||
XM_008772317.1 | XP_008770539.1 | 5 - 6 | 828 | 109 | ||
GenBank mRNA | BC158749.1 | - | 6 - 7 | 888 | 9101502 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |