Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031828.1 | NP_114016.1 | 7 - 8 | 1460 | 135 | |
GenBank mRNA | AF083341.1 | - | 7 - 8 | 896 | 135 | |
AF135265.1 | - | 7 - 8 | 1314 | 135 | ||
AY330290.1 | - | 7 - 8 | 785 | 135 | ||
AY330291.1 | - | 7 - 8 | 785 | 135 | ||
AY330292.1 | - | 7 - 8 | 785 | 135 | ||
AY330293.2 | - | 7 - 8 | 785 | 135 | ||
AY330294.1 | - | 7 - 8 | 785 | 135 | ||
AY344964.1 | - | 7 - 8 | 785 | 135 | ||
AY344965.2 | - | 7 - 8 | 1289 | 135 | ||
U40603.1 | - | 7 - 8 | 1460 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |