Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031828.1 | NP_114016.1 | 9 - 10 | 1721 | 98 | |
GenBank mRNA | AB248960.1 | - | 2 - 3 | 137 | 98 | |
AF083341.1 | - | 9 - 10 | 1157 | 98 | ||
AF135265.1 | - | 9 - 10 | 1575 | 98 | ||
AY330290.1 | - | 9 - 10 | 1046 | 98 | ||
AY330291.1 | - | 9 - 10 | 1046 | 98 | ||
AY330292.1 | - | 9 - 10 | 1055 | 98 | ||
AY330293.2 | - | 9 - 10 | 1046 | 98 | ||
AY330294.1 | - | 9 - 10 | 1046 | 98 | ||
AY344964.1 | - | 9 - 10 | 1046 | 98 | ||
U40603.1 | - | 9 - 10 | 1721 | 98 | ||
U55995.1 | - | 9 - 10 | 1199 | 98 | ||
U93052.1 | - | 9 - 10 | 1147 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |