Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175584.1 | NP_783174.1 | 1 | 985 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |