Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106097.2 | NP_001099567.2 | 3 - 4 | 348 | 135 | |
XM_006227769.2 | XP_006227831.1 | 4 - 5 | 664 | 135 | ||
GenBank mRNA | CB582092.1 | - | 3 - 4 | 348 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |