Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109136.1 | NP_001102606.1 | 1 - 2 | 330 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |