Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hey2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130417.1 | NP_569101.1 | 1 - 2 | 148 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |