Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107333.1 | NP_001100803.1 | 1 - 2 | 172 | 96 | |
XM_006227782.2 | XP_006227844.1 | 1 - 2 | 559 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |