Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024285.1 | NP_001019456.1 | 6 - 7 | 645 | 61 | |
GenBank mRNA | AB174900.2 | - | 6 - 7 | 645 | 61 | |
AY367561.1 | - | 6 - 7 | 645 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |