Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021869.2 | NP_068641.2 | 4 - 5 | 347 | 156 | |
XM_006227691.2 | XP_006227753.1 | 5 - 6 | 1669 | 156 | ||
XM_008758626.1 | XP_008756848.1 | 5 - 6 | 1663 | 156 | ||
GenBank mRNA | AF031430.1 | - | 3 - 4 | 251 | 156 | |
BC085737.1 | - | 4 - 5 | 347 | 7193349 | 156 |
Gene Symbol: LOC100910446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758670.1 | XP_008756892.1 | 2 - 3 | 233 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |