Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227800.2 | XP_006227862.1 | 11 - 12 | 1652 | 64 | |
XM_006227801.2 | XP_006227863.1 | 11 - 12 | 1589 | 64 | ||
XM_008758697.1 | XP_008756919.1 | 11 - 12 | 1725 | 64 | ||
XM_217737.8 | XP_217737.4 | 11 - 12 | 1831 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |