Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227800.2 | XP_006227862.1 | 10 - 11 | 1536 | 80 | |
XM_006227801.2 | XP_006227863.1 | 10 - 11 | 1473 | 80 | ||
XM_008758697.1 | XP_008756919.1 | 10 - 11 | 1609 | 80 | ||
XM_217737.8 | XP_217737.4 | 10 - 11 | 1715 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |