Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130428.1 | NP_569112.1 | 4 - 5 | 472 | 75 | |
GenBank mRNA | AB072907.1 | - | 4 - 5 | 472 | 75 | |
DQ402976.1 | - | 2 - 3 | 281 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |