Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033690.1 | NP_001028862.1 | 2 - 3 | 473 | 147 | |
GenBank mRNA | BC101871.1 | - | 2 - 3 | 473 | 5621693 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |