Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255191.2 | XP_006255253.1 | 1 - 2 | 979 | 60 | |
XM_006255192.2 | XP_006255254.1 | 1 - 2 | 979 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |