Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144851.1 | NP_001138323.1 | 1 - 2 | 458 | 100 | |
XM_006254007.2 | XP_006254069.1 | 1 - 2 | 395 | 100 | ||
XM_008771701.1 | XP_008769923.1 | 1 - 2 | 394 | 100 | ||
XM_008771702.1 | XP_008769924.1 | 1 - 2 | 394 | 100 | ||
GenBank mRNA | FQ212048.1 | - | 1 - 2 | 292 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |