Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107359.1 | NP_001100829.1 | 2 - 3 | 347 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |