Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759293.1 | XP_008757515.1 | 71 - 72 | 10857 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |