Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012600.2 | NP_036732.2 | 3 - 4 | 359 | 84 | |
XM_008766439.1 | XP_008764661.1 | 3 - 4 | 656 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |