Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019346.2 | NP_062219.2 | 6 - 7 | 925 | 93 | |
XM_008772168.1 | XP_008770390.1 | 7 - 8 | 1143 | 93 | ||
XM_008772169.1 | XP_008770391.1 | 6 - 7 | 2057 | 93 | ||
XM_008772170.1 | XP_008770392.1 | 6 - 7 | 994 | 93 | ||
GenBank mRNA | FQ211495.1 | - | 6 - 7 | 958 | 93 | |
X98399.1 | - | 6 - 7 | 925 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |