Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228288.2 | XP_006228350.1 | 1 - 2 | 114 | 71 | |
GenBank mRNA | FQ209745.1 | - | 1 - 2 | 41 | 71 | |
FQ212339.1 | - | 1 - 2 | 42 | 71 | ||
FQ217837.1 | - | 1 - 2 | 43 | 71 | ||
FQ221737.1 | - | 1 - 2 | 38 | 71 | ||
FQ222287.1 | - | 1 - 2 | 38 | 71 | ||
FQ223241.1 | - | 1 - 2 | 37 | 71 | ||
FQ223819.1 | - | 1 - 2 | 38 | 71 | ||
FQ224137.1 | - | 1 - 2 | 42 | 71 | ||
FQ227480.1 | - | 1 - 2 | 71 | |||
FQ228328.1 | - | 1 - 2 | 46 | 71 | ||
FQ228578.1 | - | 1 - 2 | 41 | 71 | ||
X52619.1 | - | 1 - 2 | 37 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |