Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024157.1 | NP_077071.1 | 9 - 10 | 1006 | 142 | |
XM_008761526.1 | XP_008759748.1 | 10 - 11 | 1344 | 142 | ||
GenBank mRNA | BC089798.1 | - | 9 - 10 | 1011 | 7321559 | 142 |
Y18965.1 | - | 9 - 10 | 1006 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |