Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031132.3 | NP_112394.3 | 5 - 6 | 1647 | 149 | |
XM_008766690.1 | XP_008764912.1 | 6 - 7 | 2203 | 149 | ||
GenBank mRNA | GU067683.1 | - | 5 - 6 | 1396 | 149 | |
L09653.1 | - | 5 - 6 | 1454 | 149 | ||
S67770.1 | - | 5 - 6 | 1647 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |