Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031666.1 | NP_113854.1 | 5 - 6 | 1376 | 67 | |
GenBank mRNA | AF375463.1 | - | 5 - 6 | 1376 | 67 | |
U85513.1 | - | 5 - 6 | 1508 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |