Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031743.2 | NP_113931.1 | 8 - 9 | 1739 | 100 | |
GenBank mRNA | AF021923.1 | - | 8 - 9 | 1739 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |