Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053504.1 | NP_445956.1 | 8 - 9 | 1757 | 80 | |
GenBank mRNA | AJ311671.2 | - | 8 - 9 | 1757 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |