Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053582.2 | NP_446034.1 | 7 - 8 | 1045 | 112 | |
XM_006238977.2 | XP_006239039.1 | 8 - 9 | 1137 | 112 | ||
XM_006238978.2 | XP_006239040.1 | 8 - 9 | 1134 | 112 | ||
GenBank mRNA | AB050717.1 | - | 7 - 8 | 1046 | 112 | |
BC097299.1 | - | 8 - 9 | 1104 | 7445067 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |