Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053965.2 | NP_446417.2 | 8 - 9 | 942 | 143 | |
GenBank mRNA | BC081749.1 | - | 8 - 9 | 942 | 7134670 | 143 |
FQ209681.1 | - | 8 - 9 | 898 | 143 | ||
X97831.1 | - | 8 - 9 | 899 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |