Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133405.2 | NP_596896.2 | 9 - 10 | 1542 | 81 | |
GenBank mRNA | AY033877.1 | - | 9 - 10 | 1552 | 81 | |
BC166416.1 | - | 9 - 10 | 1542 | 9033484 | 81 | |
FQ211347.1 | - | 9 - 10 | 1552 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |