Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175838.1 | NP_787032.1 | 1 - 2 | 43 | 60 | |
GenBank mRNA | BC063162.1 | - | 1 - 2 | 39 | 6918979 | 60 |
BC072542.1 | - | 1 - 2 | 38 | 7103726 | 60 | |
X61043.1 | - | 1 - 2 | 43 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |