Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198744.2 | NP_942039.1 | 4 - 5 | 524 | 82 | |
XM_006235754.2 | XP_006235816.1 | 4 - 5 | 523 | 82 | ||
XM_006235759.2 | XP_006235821.1 | 4 - 5 | 516 | 82 | ||
XM_008762499.1 | XP_008760721.1 | 4 - 5 | 457 | 82 | ||
GenBank mRNA | BC059167.1 | - | 4 - 5 | 525 | 6890828 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |