Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198778.2 | NP_942073.2 | 3 - 4 | 462 | 92 | |
XM_006234680.2 | XP_006234742.1 | 3 - 4 | 1837 | 92 | ||
XM_006234681.2 | XP_006234743.1 | 3 - 4 | 539 | 92 | ||
XM_006234682.2 | XP_006234744.1 | 3 - 4 | 626 | 92 | ||
XM_006234683.2 | XP_006234745.1 | 3 - 4 | 541 | 92 | ||
XM_008762104.1 | XP_008760326.1 | 3 - 4 | 1261 | 92 | ||
XM_008762105.1 | XP_008760327.1 | 2 - 3 | 1521 | 92 | ||
GenBank mRNA | AY093422.1 | - | 3 - 4 | 512 | 92 | |
BC161828.1 | - | 3 - 4 | 544 | 9033590 | 92 | |
EV763409.1 | - | 3 - 4 | 566 | 92 | ||
EV768250.1 | - | 1 - 2 | 97 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |