Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpgt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199494.1 | NP_955788.1 | 3 - 4 | 347 | 145 | |
GenBank mRNA | AJ276066.1 | - | 3 - 4 | 382 | 145 | |
BC168244.1 | - | 3 - 4 | 330 | 5600080 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |