Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201419.1 | NP_958822.1 | 13 - 14 | 2245 | 96 | |
GenBank mRNA | AY333961.1 | - | 13 - 14 | 2245 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |