Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113335.1 | NP_001106806.1 | 10 - 11 | 2164 | 149 | |
NM_012653.2 | NP_036785.2 | 9 - 10 | 2161 | 149 | ||
GenBank mRNA | L11004.1 | - | 9 - 10 | 2176 | 149 | |
L11236.1 | - | 10 - 11 | 2164 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |