Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013033.2 | NP_037165.1 | 7 - 8 | 836 | 78 | |
GenBank mRNA | AB000729.1 | - | 7 - 8 | 862 | 78 | |
BC081827.1 | - | 7 - 8 | 868 | 7129017 | 78 | |
D16101.1 | - | 7 - 8 | 836 | 78 | ||
U03120.1 | - | 7 - 8 | 794 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |