Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020075.1 | NP_064460.1 | 3 - 4 | 554 | 88 | |
XM_006240608.1 | XP_006240670.1 | 5 - 6 | 817 | 88 | ||
XM_006240609.2 | XP_006240671.1 | 4 - 5 | 1097 | 88 | ||
GenBank mRNA | BC062398.1 | - | 4 - 5 | 742 | 5623373 | 88 |
FQ222051.1 | - | 1 - 2 | 634 | 88 | ||
L11651.1 | - | 3 - 4 | 554 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |