Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022546.1 | NP_071991.1 | 5 - 6 | 715 | 84 | |
XM_006240991.1 | XP_006241053.1 | 5 - 6 | 846 | 84 | ||
XM_006240993.2 | XP_006241055.1 | 5 - 6 | 953 | 84 | ||
XM_008765129.1 | XP_008763351.1 | 5 - 6 | 1179 | 84 | ||
GenBank mRNA | AJ006971.1 | - | 5 - 6 | 715 | 84 | |
BC062076.1 | - | 5 - 6 | 716 | 5599435 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |